missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-2,6-dichloropyridine, 97%

Catalog No. p-7030750
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3159806 5 g
AAH3159814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3159806 Supplier Thermo Scientific Chemicals Supplier No. H3159806

CAS: 62476-56-6 | C5H4Cl2N2 | 163.00 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 62476-56-6
Molecular Formula C5H4Cl2N2
Molecular Weight (g/mol) 163.00
MDL Number MFCD00023417
InChI Key MJVZSRZTBDMYLX-UHFFFAOYSA-N
Synonym 3-amino-2,6-dichloropyridine, 2,6-dichloro-3-aminopyridine, 2,6-dichloro-3-pyridylamine, 3-pyridinamine, 2,6-dichloro, 2,6-dichloro-3-pyridinamine, 2,6-dichloropyridin-3-ylamine, 2,6-dichloro-pyridin-3-ylamine, 2,6-dichlorpyridin-3-amin, pubchem10384, ksc495k2f
PubChem CID 112846
IUPAC Name 2,6-dichloropyridin-3-amine
SMILES NC1=CC=C(Cl)N=C1Cl

Specifications

Melting Point 120°C to 124°C
Quantity 5 g
Formula Weight 163.01
Percent Purity 97%
Chemical Name or Material 3-Amino-2,6-dichloropyridine
Hazard Category H315-H319
Hazard Statement GHS H Statement
H301-H311-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
EINECSNumber 263-559-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only