missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-2,6-dibromopyridine 98.0+%, TCI America™

Catalog No. p-7117818
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
Catalog No. Quantity
A22721G 1 g
A22725G 5 g
2 options

Catalog No. A22721G

Supplier: TCI America A22721G

Only null left

Chemical Identifiers

CAS 39856-57-0
Molecular Formula C5H4Br2N2
Molecular Weight (g/mol) 251.909
MDL Number MFCD00128862
InChI Key CRTOIQFRVBJJRI-UHFFFAOYSA-N
Synonym 3-amino-2,6-dibromopyridine, 2,6-dibromo-3-pyridinamine, 2,6-dibromo-pyridin-3-ylamine, 3-pyridinamine, 2,6-dibromo, 2,6-dibromo-3-pyridylamine, 2,6-dibromo-3-aminopyridine, zlchem 949, pubchem9809, acmc-209j8e, ksc495i3f
PubChem CID 817678
IUPAC Name 2,6-dibromopyridin-3-amine
SMILES C1=CC(=NC(=C1N)Br)Br

Specifications

Melting Point 147°C
Quantity 1 g
Formula Weight 251.91
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Amino-2,6-dibromopyridine