missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-2,4-dimethylpyridine, 97%

Catalog No. AAH6673206
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6673206 5 g
AAH6673203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6673206 Supplier Thermo Scientific Chemicals Supplier No. H6673206
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1073-21-8 | C7H10N2 | 122.171 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1073-21-8
Molecular Formula C7H10N2
Molecular Weight (g/mol) 122.171
MDL Number MFCD08235192
InChI Key FSYHJEBPLDNXAM-UHFFFAOYSA-N
PubChem CID 14910781
IUPAC Name 2,4-dimethylpyridin-3-amine
SMILES CC1=C(C(=NC=C1)C)N

Specifications

Color Brown
Quantity 5 g
UN Number UN2811
Beilstein 111999
Synonym 3-amino-2,4-dimethylpyridine, 3-amino-2,4-lutidine, 3-pyridinamine, 2,4-dimethyl, 2,4-dimethyl-3-pyridylamine, 2,4-dimethyl-3-pyridinamine, 2,4-dimethyl-3-aminopyridine, 3-pyridinamine,2,4-dimethyl, 2,4-dimethyl-pyridin-3-ylamine, 2,4-dimethylpyridin-3-amine, 3-amino-2,4-lutidine
Formula Weight 122.17
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 3-Amino-2,4-dimethylpyridine
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only