missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-2,4-difluorobenzeneboronic acid pinacol ester, 96%

Catalog No. AAH59033MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH59033MD 250 mg
AAH5903303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH59033MD Supplier Thermo Scientific Chemicals Supplier No. H59033MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1315692-91-1 | C12H16BF2NO2 | 255.072 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1315692-91-1
Molecular Formula C12H16BF2NO2
Molecular Weight (g/mol) 255.072
MDL Number MFCD22419270
InChI Key MQGLBRAIKSVSIT-UHFFFAOYSA-N
Synonym 3-amino-2,4-difluorobenzeneboronic acid pinacol ester, 2,6-difluoro-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, 3-amino-2,4-difluorobenzeneboronicacidpinacolester, 2,6-difluoro-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl aniline
PubChem CID 99738339
IUPAC Name 2,6-difluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
SMILES B1(OC(C(O1)(C)C)(C)C)C2=C(C(=C(C=C2)F)N)F

Specifications

Melting Point 75°C to 77°C
Quantity 250 mg
Formula Weight 255.07
Percent Purity 96%
Chemical Name or Material 3-Amino-2,4-difluorobenzeneboronic acid pinacol ester
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only