missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 3'-Amino-2',3'-dideoxythymidine, 99%

Catalog No. AAJ6434203
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6434203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ6434203 Supplier Thermo Scientific Chemicals Supplier No. J6434203

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 52450-18-7 | C10H15N3O4 | 241.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 52450-18-7
Molecular Formula C10H15N3O4
Molecular Weight (g/mol) 241.25
MDL Number MFCD00038057
InChI Key ADVCGXWUUOVPPB-XLPZGREQSA-N
Synonym 3'-amino-3'-deoxythymidine, 3'-amino-2',3'-dideoxythymidine, unii-7w21m0c25b, 3'-deoxy-3'-aminothymidine, 3'-amino-3'-deoxy-d-thymidine, thymidine, 3'-amino-3'-deoxy, 1-2r,4s,5s-4-amino-5-hydroxymethyl oxolan-2-yl-5-methylpyrimidine-2,4-dione, 1-2r,4s,5s-4-amino-5-hydroxymethyl tetrahydrofuran-2-yl-5-methyl-pyrimidine-2,4-dione, 1-2r,4s,5s-4-amino-5-hydroxymethyl tetrahydrofuran-2-yl-5-methylpyrimidine-2,4 1h,3h-dione, 1-2r,4s,5s-4-amino-5-hydroxymethyl oxolan-2-yl-5-methyl-3h-pyrimidine-2,4-dione
PubChem CID 108074
IUPAC Name 1-[(2R,4S,5S)-4-amino-5-(hydroxymethyl)oxolan-2-yl]-5-methylpyrimidine-2,4-dione
SMILES CC1=CN([C@H]2C[C@H](N)[C@@H](CO)O2)C(=O)NC1=O

Specifications

Color White to Yellow
Quantity 1 g
Formula Weight 241.24
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 3'-Amino-2',3'-dideoxythymidine
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only