missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-1H-pyrazole-4-carbonitrile, 98%

Catalog No. AAA1426614
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1426614 25 g
AAA1426606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1426614 Supplier Thermo Scientific Chemicals Supplier No. A1426614
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 16617-46-2 | C4H4N4 | 108.104 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 16617-46-2
Molecular Formula C4H4N4
Molecular Weight (g/mol) 108.104
MDL Number MFCD00005237
InChI Key FFNKBQRKZRMYCL-UHFFFAOYSA-N
Synonym 3-amino-4-pyrazolecarbonitrile, 3-amino-1h-pyrazole-4-carbonitrile, 3-amino-4-cyanopyrazole, 3-aminopyrazole-4-carbonitrile, 5-amino-4-cyanopyrazole, 1h-pyrazole-4-carbonitrile, 3-amino, 5-aminopyrazole-4-carbonitrile, 3-amino-4-cyano-1h-pyrazole, 4-cyano-5-aminopyrazole, 3-amino-4-cyano pyrazole
PubChem CID 85515
IUPAC Name 5-amino-1H-pyrazole-4-carbonitrile
SMILES C1=NNC(=C1C#N)N

Specifications

Melting Point 173°C to 176°C
Odor Odorless
Quantity 25 g
UN Number UN3439
Beilstein 2647
Formula Weight 108.1
Percent Purity 98%
Chemical Name or Material 3-Amino-1H-pyrazole-4-carbonitrile
Hazard Category H302+H312+H332-H315-H319
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
EINECSNumber 240-665-6
TSCA Yes

RUO – Research Use Only