missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-1H-indazole, 97%

Catalog No. p-7032093
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3343503 1 g
AAH3343509 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3343503 Supplier Thermo Scientific Chemicals Supplier No. H3343503

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 874-05-5 | C7H7N3 | 133.154 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 874-05-5
Molecular Formula C7H7N3
Molecular Weight (g/mol) 133.154
MDL Number MFCD00182045
InChI Key YDTDKKULPWTHRV-UHFFFAOYSA-N
Synonym 3-amino-1h-indazole, 1h-indazol-3-ylamine, 3-aminoindazole, 3-indazolamine, 2h-indazol-3-amine, 1h-indazole, 3-amino, aminoindazole, amino-indazole, 1h-indazole-3-ylamine, indazole amine
PubChem CID 13399
IUPAC Name 1H-indazol-3-amine
SMILES C1=CC=C2C(=C1)C(=NN2)N

Specifications

Melting Point 150°C to 152°C
Quantity 1 g
UN Number UN2811
Formula Weight 133.15
Percent Purity 97%
Chemical Name or Material 3-Amino-1H-indazole
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only