missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-1H-1,2,4-triazole-5-carboxylic acid hydrate, 98%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 249301000

 View more versions of this product

Catalog No. AC249301000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

Specifications

Specifications

3-Amino-1H-1, 2, 4-triazole-5-carboxylic acid hydrate
2% max.
98%
C3H4N4O2·xH2O
100g
Solubility in water: .. Other solubilities: insoluble in dil. acid- soluble in aq. alkalis
128.09
1266615-63-7,3641-13-2
Authentic
Glass bottle
MFCD00149595
5-Amino-2H-1, 2, 4-triazole-3-carboxylic acid hydrate
128.09
98%

Chemical Identifiers

1266615-63-7
128.09
5-Amino-2H-1, 2, 4-triazole-3-carboxylic acid hydrate
C3H4N4O2·xH2O
MFCD00149595
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only