missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-1-propanol 99.0+%, TCI America™

Supplier:  TCI America A0438500G

 View more versions of this product

Catalog No. A0438500G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

3-Amino-1-propanol
Yellow
C3H9NO
500 g
3-aminopropanol, 3-amino-1-propanol, propanolamine, 1-propanol, 3-amino, 3-propanolamine, 1-amino-3-propanol, 3-hydroxypropylamine, 3-aminopropyl alcohol, 1,3-propanolamine, beta-alaninol
C(CN)CO
75.111
75.11
Liquid
156-87-6
188°C
MFCD00008223
2735
WUGQZFFCHPXWKQ-UHFFFAOYSA-N
3-aminopropan-1-ol
9086
≥99.0% (GC,T)

Chemical Identifiers

156-87-6
75.111
WUGQZFFCHPXWKQ-UHFFFAOYSA-N
9086
C(CN)CO
C3H9NO
MFCD00008223
3-aminopropanol, 3-amino-1-propanol, propanolamine, 1-propanol, 3-amino, 3-propanolamine, 1-amino-3-propanol, 3-hydroxypropylamine, 3-aminopropyl alcohol, 1,3-propanolamine, beta-alaninol
3-aminopropan-1-ol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0323

RTECSNumber : UA5600000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions