missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-1-propanol 99.0+%, TCI America™

Catalog No. A043825G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A043825G 25 g
A0438500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A043825G Supplier TCI America Supplier No. A043825G

Chemical Identifiers

CAS 156-87-6
Molecular Formula C3H9NO
Molecular Weight (g/mol) 75.111
MDL Number MFCD00008223
InChI Key WUGQZFFCHPXWKQ-UHFFFAOYSA-N
Synonym 3-aminopropanol, 3-amino-1-propanol, propanolamine, 1-propanol, 3-amino, 3-propanolamine, 1-amino-3-propanol, 3-hydroxypropylamine, 3-aminopropyl alcohol, 1,3-propanolamine, beta-alaninol
PubChem CID 9086
IUPAC Name 3-aminopropan-1-ol
SMILES C(CN)CO

Specifications

Color Yellow
Boiling Point 188°C
Quantity 25 g
UN Number 2735
Formula Weight 75.11
Percent Purity ≥99.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 3-Amino-1-propanol
EINECSNumber (2)-0323
RTECSNumber UA5600000
TSCA Yes