missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-1-propanol, 99%

Catalog No. AC104460050
Change view
Click to view available options
Quantity:
5 mL
500 mL
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC104460050 5 mL Glass bottle
AC104465000 500 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC104460050 Supplier Thermo Scientific Chemicals Supplier No. 104460050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 156-87-6 | C3H9NO | 75.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 156-87-6
MDL Number MFCD00008223
InChI Key WUGQZFFCHPXWKQ-UHFFFAOYSA-N
Synonym 3-aminopropanol, 3-amino-1-propanol, propanolamine, 1-propanol, 3-amino, 3-propanolamine, 1-amino-3-propanol, 3-hydroxypropylamine, 3-aminopropyl alcohol, 1,3-propanolamine, beta-alaninol
PubChem CID 9086
IUPAC Name 3-aminopropan-1-ol
SMILES C(CN)CO

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 10.0°C to 12.0°C
Color Colorless to Yellow
Density 0.9820g/mL
Boiling Point 187.0°C to 188.0°C
Flash Point 101°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Molecular Formula C3H9NO
Refractive Index 1.4595 to 1.4615
Linear Formula H2N(CH2)3OH
Quantity 5 mL
Beilstein 04, 288
Specific Gravity 0.982
Solubility Information Solubility in water: freely soluble. Other solubilities: miscible with alcohols,esters, chloroform, acetone, slightly soluble in arom. and aliph. hydrocarbons
Molecular Weight (g/mol) 75.11
Viscosity 36 mPa.s (20°C)
Formula Weight 75.11
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 3-Amino-1-propanol, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water
EINECSNumber 205-864-4
RTECSNumber UA5600000
TSCA TSCA

RUO – Research Use Only