missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-1-propanesulfonic acid, 95%

Catalog No. AC104430010
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC104430010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC104430010 Supplier Thermo Scientific Chemicals Supplier No. 104430010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 3687-18-1 | C3H9NO3S | 139.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3687-18-1
Molecular Formula C3H9NO3S
Molecular Weight (g/mol) 139.17
MDL Number MFCD00008225
InChI Key SNKZJIOFVMKAOJ-UHFFFAOYSA-N
Synonym homotaurine, 3-amino-1-propanesulfonic acid, tramiprosate, alzhemed, 3-aminopropanesulphonic acid, 3aps, 1-propanesulfonic acid, 3-amino, cerebril, 3-aminopropanesulfonic acid, 3-aminopropane sulfonic acid
PubChem CID 1646
ChEBI CHEBI:1457
IUPAC Name 3-aminopropane-1-sulfonic acid
SMILES NCCCS(O)(=O)=O

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point >270°C
Color Blue to White
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Linear Formula H2N(CH2)3SO3H
Quantity 1 g
Merck Index 15, 9728
Formula Weight 139.17
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material 3-Amino-1-propanesulfonic acid
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER
EINECSNumber 222-977-4

RUO – Research Use Only