missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-1-phenylbutane 98.0+%, TCI America™

Catalog No. p-7117178
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A099925ML 25 mL
A0999500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A099925ML Supplier TCI America Supplier No. A099925ML

Chemical Identifiers

CAS 22374-89-6
Molecular Formula C10H15N
Molecular Weight (g/mol) 149.237
MDL Number MFCD00008090
InChI Key WECUIGDEWBNQJJ-UHFFFAOYSA-N
Synonym 1-methyl-3-phenylpropylamine, 3-amino-1-phenylbutane, 2-amino-4-phenylbutane, 1-phenyl-3-aminobutane, 4-phenyl-2-aminobutane, 4-phenyl-2-butylamine, 1-phenyl-3-amino-butan, benzenepropanamine, .alpha.-methyl, alpha-methylbenzenepropanamine, propylamine, 1-methyl-3-phenyl
PubChem CID 31160
IUPAC Name 4-phenylbutan-2-amine
SMILES CC(CCC1=CC=CC=C1)N

Specifications

Color Colorless
Boiling Point 222°C
Quantity 25 mL
UN Number 2735
Formula Weight 149.24
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 3-Amino-1-phenylbutane
RTECSNumber UI3920000
TSCA No