missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-1-phenyl-2-pyrazolin-5-one 98.0+%, TCI America™

Catalog No. A103425G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A103425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A103425G Supplier TCI America Supplier No. A103425G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4149-06-8
Molecular Formula C9H9N3O
Molecular Weight (g/mol) 175.19
MDL Number MFCD00003137
InChI Key LPOVZHYARSAVIZ-UHFFFAOYSA-N
Synonym 3-amino-1-phenyl-2-pyrazolin-5-one, 3-amino-1-phenyl-1h-pyrazol-5 4h-one, 3h-pyrazol-3-one, 5-amino-2,4-dihydro-2-phenyl, 3-amino-1-phenyl-4,5-dihydro-1h-pyrazol-5-one, 5-pyrazolone, 3-amino-1-phenyl, 5-amino-2-phenyl-2,4-dihydro-3h-pyrazol-3-one, 3h-pyrazol-3-one, 2,4-dihydro-5-amino-2-phenyl, 2,4-dihydro-5-amino-2-phenyl-3h-pyrazol-3-one, acmc-1arey, lpovzhyarsaviz-uhfffaoysa
PubChem CID 77794
IUPAC Name 5-amino-2-phenyl-4H-pyrazol-3-one
SMILES C1C(=NN(C1=O)C2=CC=CC=C2)N

Specifications

Melting Point 206°C
Quantity 25 g
Formula Weight 175.19
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Amino-1-phenyl-2-pyrazolin-5-one
EINECSNumber (5)-0286
RTECSNumber UR0393500
TSCA No