missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-1-phenyl-2-pyrazolin-5-one, 97%

Catalog No. AAA1086222
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1086222 100 g
AAA1086214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1086222 Supplier Thermo Scientific Chemicals Supplier No. A1086222
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 4149-06-8 | C9H9N3O | 175.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4149-06-8
Molecular Formula C9H9N3O
Molecular Weight (g/mol) 175.19
MDL Number MFCD00003137
InChI Key LPOVZHYARSAVIZ-UHFFFAOYSA-N
Synonym 3-amino-1-phenyl-2-pyrazolin-5-one, 3-amino-1-phenyl-1h-pyrazol-5 4h-one, 3h-pyrazol-3-one, 5-amino-2,4-dihydro-2-phenyl, 3-amino-1-phenyl-4,5-dihydro-1h-pyrazol-5-one, 5-pyrazolone, 3-amino-1-phenyl, 5-amino-2-phenyl-2,4-dihydro-3h-pyrazol-3-one, 3h-pyrazol-3-one, 2,4-dihydro-5-amino-2-phenyl, 2,4-dihydro-5-amino-2-phenyl-3h-pyrazol-3-one, acmc-1arey, lpovzhyarsaviz-uhfffaoysa
PubChem CID 77794
IUPAC Name 5-amino-2-phenyl-4H-pyrazol-3-one
SMILES C1C(=NN(C1=O)C2=CC=CC=C2)N

Specifications

Melting Point ∼213°C (decomposition)
Quantity 100 g
Beilstein 153681
Formula Weight 175.19
Percent Purity 97%
Chemical Name or Material 3-Amino-1-phenyl-2-pyrazolin-5-one
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 223-974-0
RTECSNumber UR0393500
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only