missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-1-adamantanol, 98+%

Catalog No. AAH6413706
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6413706 5 g
AAH6413714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6413706 Supplier Thermo Scientific Chemicals Supplier No. H6413706

CAS: 702-82-9 | C10H17NO | 167.252 g/mol

It is employed in the synthesis of inhibitors with antihyperglycemic properties. vil- dagliptin is synthesised from 3-Amino-1-adamantanol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed in the synthesis of inhibitors with antihyperglycemic properties. vil- dagliptin is synthesised from 3-Amino-1-adamantanol.

Solubility
Soluble in organic solvents,insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 702-82-9
Molecular Formula C10H17NO
Molecular Weight (g/mol) 167.252
MDL Number MFCD01821204
InChI Key DWPIPTNBOVJYAD-UHFFFAOYSA-N
Synonym 3-amino-1-adamantanol, 3-amino-1-hydroxyadamantane, 1-amino-3-adamantanol, 3-amino-adamantanol, 1-aminoadamantane-3-ol, 3-hydroxy-1-aminoadamantane, 1-amino-3-hydroxyadamantane, tricyclo 3.3.1.13,7 decan-1-ol, 3-amino, 3-azanyladamantan-1-ol, 3-hydroxyadamantane amine
PubChem CID 658645
IUPAC Name 3-aminoadamantan-1-ol
SMILES C1C2CC3(CC1CC(C2)(C3)O)N

Specifications

Melting Point 265°C (literature)
Flash Point >110°C (230°F)
Quantity 5 g
Beilstein 2802127
Solubility Information Soluble in organic solvents,insoluble in water.
Formula Weight 167.25
Percent Purity ≥98%
Physical Form Solid
Chemical Name or Material 3-Amino-1-adamantanol
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only