missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-1,2-propanediol 98.0+%, TCI America™

Catalog No. A062825G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A062825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A062825G Supplier TCI America Supplier No. A062825G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 616-30-8
Color Yellow
Molecular Formula C3H9NO2
Quantity 25 g
MDL Number MFCD00008140
UN Number 2735
Synonym 3-amino-1,2-propanediol, 2,3-dihydroxypropylamine, 1,2-propanediol, 3-amino, 1-amino-2,3-propanediol, 1-aminoglycerol, 2,3-propandiol-1-amine, 1-aminopropanediol, 3-aminopropan-1,2-diol, 3-amino-1,2-proopanediol, +/--3-amino-1,2-propanediol
InChI Key KQIGMPWTAHJUMN-UHFFFAOYSA-N
SMILES C(C(CO)O)N
IUPAC Name 3-aminopropane-1,2-diol
Molecular Weight (g/mol) 91.11
PubChem CID 73561
Formula Weight 91.11
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 3-Amino-1,2-propanediol
Show More Show Less
EINECSNumber (2)-3504
RTECSNumber TY2800000
TSCA Yes