missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-1,2-propanediol, 98%

Catalog No. AC104420250
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC104420250 25 g Glass bottle
AC104421000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC104420250 Supplier Thermo Scientific Chemicals Supplier No. 104420250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 616-30-8 | C3H9NO2 | 91.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 616-30-8
Molecular Weight (g/mol) 91.11
MDL Number MFCD00008140
InChI Key KQIGMPWTAHJUMN-UHFFFAOYSA-N
Synonym 3-amino-1,2-propanediol, 2,3-dihydroxypropylamine, 1,2-propanediol, 3-amino, 1-amino-2,3-propanediol, 1-aminoglycerol, 2,3-propandiol-1-amine, 1-aminopropanediol, 3-aminopropan-1,2-diol, 3-amino-1,2-proopanediol, +/--3-amino-1,2-propanediol
PubChem CID 73561
IUPAC Name 3-aminopropane-1,2-diol
SMILES C(C(CO)O)N

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Density 1.1700g/mL
Boiling Point 264.0°C to 265.0°C (739.0 mmHg)
Flash Point 155°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C3H9NO2
Refractive Index 1.4900 to 1.4940
Linear Formula H2NCH2CH(OH)CH2OH
Quantity 25 g
Beilstein 04,301
Specific Gravity 1.17
Solubility Information Solubility in water: soluble in water. Other solubilities: soluble in alcohols
Viscosity 3 Pas (20°C)
Formula Weight 91.11
Percent Purity 98%
Physical Form Viscous Liquid
Chemical Name or Material 3-Amino-1, 2-propanediol, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 210-475-8
RTECSNumber TY2800000
TSCA TSCA

RUO – Research Use Only