missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-3-Amino-1,2-propanediol, 97%

Catalog No. p-7026598
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2267214 25 g
AAB2267222 100 g
AAB2267236 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2267214 Supplier Thermo Scientific Chemicals Supplier No. B2267214

CAS: 616-30-8 | C3H9NO2 | 91.11 g/mol

3-Amino-1,2-propanediol is used in the preparation of iohexol, which is a X-CT non-ionic contrast medium. It acts as a starting material in the production of specialty materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Amino-1,2-propanediol is used in the preparation of iohexol, which is a X-CT non-ionic contrast medium. It acts as a starting material in the production of specialty materials.

Solubility
Miscible with water.

Notes
Hygroscopic. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 616-30-8
Molecular Formula C3H9NO2
Molecular Weight (g/mol) 91.11
MDL Number MFCD00008140
InChI Key KQIGMPWTAHJUMN-UHFFFAOYSA-N
Synonym 3-amino-1,2-propanediol, 2,3-dihydroxypropylamine, 1,2-propanediol, 3-amino, 1-amino-2,3-propanediol, 1-aminoglycerol, 2,3-propandiol-1-amine, 1-aminopropanediol, 3-aminopropan-1,2-diol, 3-amino-1,2-proopanediol, +/--3-amino-1,2-propanediol
PubChem CID 73561
IUPAC Name 3-aminopropane-1,2-diol
SMILES C(C(CO)O)N

Specifications

Density 1.186
Boiling Point 264°C to 266°C (739 mmHg)
Flash Point 186°C (366°F)
Refractive Index 1.492
Quantity 25 g
UN Number UN2735
Beilstein 1719121
Sensitivity Hygroscopic
Solubility Information Miscible with water.
Formula Weight 91.11
Percent Purity 97%
Chemical Name or Material (+/-)-3-Amino-1,2-propanediol
Show More Show Less
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
EINECSNumber 210-475-8
RTECSNumber TY2800000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only