missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-1,2,4-triazine, 97%

CAS: 1120-99-6 | C3H4N4 | 96.09 g/mol

$177.07 - $700.00

Chemical Identifiers

CAS 1120-99-6
Molecular Formula C3H4N4
Molecular Weight (g/mol) 96.09
MDL Number MFCD00006459
InChI Key MJIWQHRXSLOUJN-UHFFFAOYSA-N
Synonym 3-amino-1,2,4-triazine, as-triazine, 3-amino, 3-amino-as-triazine, 1,2,4-triazin-3-ylamine, 1,2,4-triazine-3-ylamine, 1,2,4 triazin-3-ylamine, zlchem 1277, 1,2,4-triazine-3-amine, 3-amino-1,2,4,-triazine
PubChem CID 70715
IUPAC Name 1,2,4-triazin-3-amine
SMILES C1=CN=NC(=N1)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC146230100
SDS View Documents
Thermo Scientific Chemicals
146230100
10 g Glass bottle
Each for $177.07
Only null left
 
AC146230500
SDS View Documents
Thermo Scientific Chemicals
146230500
50 g Glass bottle
Each for $700.00
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1120-99-6
96.09
MJIWQHRXSLOUJN-UHFFFAOYSA-N
70715
C1=CN=NC(=N1)N
C3H4N4
MFCD00006459
3-amino-1,2,4-triazine, as-triazine, 3-amino, 3-amino-as-triazine, 1,2,4-triazin-3-ylamine, 1,2,4-triazine-3-ylamine, 1,2,4 triazin-3-ylamine, zlchem 1277, 1,2,4-triazine-3-amine, 3-amino-1,2,4,-triazine
1,2,4-triazin-3-amine

Specifications

1120-99-6
100.0
Brown
97%
C3H4N4
10 g
MJIWQHRXSLOUJN-UHFFFAOYSA-N
1,2,4-triazin-3-amine
70715
97%
3-Amino-1, 2, 4-triazine
96.0
175°C to 177°C
Authentic
Glass bottle
MFCD00006459
3-amino-1,2,4-triazine, as-triazine, 3-amino, 3-amino-as-triazine, 1,2,4-triazin-3-ylamine, 1,2,4-triazine-3-ylamine, 1,2,4 triazin-3-ylamine, zlchem 1277, 1,2,4-triazine-3-amine, 3-amino-1,2,4,-triazine
C1=CN=NC(=N1)N
96.09
96.09
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 214-324-7

SDS
Documents

Documents

RUO – Research Use Only