missing translation for 'onlineSavingsMsg'
Learn More

3-Amidinopyridine hydrochloride, 97%

Catalog No. p-7025081
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2040603 1 g
AAB2040606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2040603 Supplier Thermo Scientific Chemicals Supplier No. B2040603
Only null left

CAS: 7356-60-7 | C6H8ClN3 | 157.601 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7356-60-7
Molecular Formula C6H8ClN3
Molecular Weight (g/mol) 157.601
MDL Number MFCD00054326
InChI Key MKJPBOVLAZADQJ-UHFFFAOYSA-N
Synonym nicotinimidamide hydrochloride, 3-amidinopyridine hydrochloride, pyridine-3-carboximidamide hydrochloride, nicotinamidine hydrochloride, 3-pyridinecarboximidamide, monohydrochloride, nicotinamidine, hydrochloride, pyridine-3-carbonamidine hydrochloride, pyridine-3-carboxamidine hydrochloride, 3-pyridinecarboxamidine hydrochloride, pyridine-3-carboximidamide hcl
PubChem CID 202042
IUPAC Name pyridine-3-carboximidamide;hydrochloride
SMILES C1=CC(=CN=C1)C(=N)N.Cl

Specifications

Melting Point 187°C to 189°C
Quantity 1 g
Sensitivity Hygroscopic
Formula Weight 157.6
Percent Purity 97%
Chemical Name or Material 3-Amidinopyridine hydrochloride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only