missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Allylrhodanine 98.0+%, TCI America™
$152.86 - $495.84
Chemical Identifiers
| CAS | 1457-47-2 |
|---|---|
| Molecular Formula | C6H7NOS2 |
| Molecular Weight (g/mol) | 173.248 |
| MDL Number | MFCD00044700 |
| InChI Key | GYGUTBCTEJBRAN-UHFFFAOYSA-N |
| Synonym | 3-allylrhodanine, rhodanine, 3-allyl, n-allylrhodanine, 3-prop-2-en-1-yl-2-sulfanylidene-1,3-thiazolidin-4-one, allyl-4-oxo-2-thioxothiazolidin, 4-thiazolidinone, 3-2-propenyl-2-thioxo, n-allyl rhodanine, 3-allyl-2-thioxo-thiazolidin-4-one, 3-allyl-2-thioxo-1,3-thiazolidin-4-one, 3-prop-2-en-1-yl-2-thioxo-1,3-thiazolidin-4-one |
| PubChem CID | 73823 |
| IUPAC Name | 3-prop-2-enyl-2-sulfanylidene-1,3-thiazolidin-4-one |
| SMILES | C=CCN1C(=O)CSC1=S |
Chemical Identifiers
| 1457-47-2 | |
| 173.248 | |
| GYGUTBCTEJBRAN-UHFFFAOYSA-N | |
| 73823 | |
| C=CCN1C(=O)CSC1=S |
| C6H7NOS2 | |
| MFCD00044700 | |
| 3-allylrhodanine, rhodanine, 3-allyl, n-allylrhodanine, 3-prop-2-en-1-yl-2-sulfanylidene-1,3-thiazolidin-4-one, allyl-4-oxo-2-thioxothiazolidin, 4-thiazolidinone, 3-2-propenyl-2-thioxo, n-allyl rhodanine, 3-allyl-2-thioxo-thiazolidin-4-one, 3-allyl-2-thioxo-1,3-thiazolidin-4-one, 3-prop-2-en-1-yl-2-thioxo-1,3-thiazolidin-4-one | |
| 3-prop-2-enyl-2-sulfanylidene-1,3-thiazolidin-4-one |
Specifications
| 1457-47-2 | |
| 139°C | |
| MFCD00044700 | |
| 3-allylrhodanine, rhodanine, 3-allyl, n-allylrhodanine, 3-prop-2-en-1-yl-2-sulfanylidene-1,3-thiazolidin-4-one, allyl-4-oxo-2-thioxothiazolidin, 4-thiazolidinone, 3-2-propenyl-2-thioxo, n-allyl rhodanine, 3-allyl-2-thioxo-thiazolidin-4-one, 3-allyl-2-thioxo-1,3-thiazolidin-4-one, 3-prop-2-en-1-yl-2-thioxo-1,3-thiazolidin-4-one | |
| C=CCN1C(=O)CSC1=S | |
| 173.248 | |
| 173.25 | |
| Crystalline Powder |
| 47°C | |
| C6H7NOS2 | |
| 5 g | |
| GYGUTBCTEJBRAN-UHFFFAOYSA-N | |
| 3-prop-2-enyl-2-sulfanylidene-1,3-thiazolidin-4-one | |
| 73823 | |
| ≥98.0% (GC) | |
| 3-Allylrhodanine |
Safety and Handling
EINECSNumber : (5)-3405
TSCA : No