missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Allyloxy-1,2-propanediol 99.0+%, TCI America™
$50.33 - $177.08
Chemical Identifiers
| CAS | 123-34-2 |
|---|---|
| Molecular Formula | C6H12O3 |
| Molecular Weight (g/mol) | 132.159 |
| MDL Number | MFCD00004721 |
| InChI Key | PAKCOSURAUIXFG-UHFFFAOYSA-N |
| Synonym | 3-allyloxy-1,2-propanediol, 3-allyloxy propane-1,2-diol, glycerol 1-allylether, ether, allyl glyceryl, 3-allyloxy propanediol, 1-allyloxy-2,3-propanediol, glycerin 1-allyl ether, glycerol alpha-allyl ether, glycerol alpha-monoallyl ether, 3-2-propenyloxy-1,2-propanediol |
| PubChem CID | 78950 |
| IUPAC Name | 3-prop-2-enoxypropane-1,2-diol |
| SMILES | C=CCOCC(CO)O |
Chemical Identifiers
| 123-34-2 | |
| 132.159 | |
| PAKCOSURAUIXFG-UHFFFAOYSA-N | |
| 78950 | |
| C=CCOCC(CO)O |
| C6H12O3 | |
| MFCD00004721 | |
| 3-allyloxy-1,2-propanediol, 3-allyloxy propane-1,2-diol, glycerol 1-allylether, ether, allyl glyceryl, 3-allyloxy propanediol, 1-allyloxy-2,3-propanediol, glycerin 1-allyl ether, glycerol alpha-allyl ether, glycerol alpha-monoallyl ether, 3-2-propenyloxy-1,2-propanediol | |
| 3-prop-2-enoxypropane-1,2-diol |
Specifications
| 123-34-2 | |
| 6.5 tp 7.5 (50% H2O 20°C) | |
| C6H12O3 | |
| 25 g | |
| PAKCOSURAUIXFG-UHFFFAOYSA-N | |
| 3-prop-2-enoxypropane-1,2-diol | |
| 78950 | |
| ≥99.0% (GC) | |
| 3-Allyloxy-1,2-propanediol |
| Yellow | |
| 142°C | |
| MFCD00004721 | |
| 3-allyloxy-1,2-propanediol, 3-allyloxy propane-1,2-diol, glycerol 1-allylether, ether, allyl glyceryl, 3-allyloxy propanediol, 1-allyloxy-2,3-propanediol, glycerin 1-allyl ether, glycerol alpha-allyl ether, glycerol alpha-monoallyl ether, 3-2-propenyloxy-1,2-propanediol | |
| C=CCOCC(CO)O | |
| 132.159 | |
| 132.16 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-0416
RTECSNumber : TY2700000
TSCA : Yes