missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Allyloxy-1,2-propanediol 99.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| pH | 6.5 tp 7.5 (50% H2O 20°C) |
| Boiling Point | 142°C |
| Quantity | 500 g |
| Formula Weight | 132.16 |
| Percent Purity | ≥99.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | 3-Allyloxy-1,2-propanediol |
Safety and Handling
| EINECSNumber | (2)-0416 |
| RTECSNumber | TY2700000 |
| TSCA | Yes |