missing translation for 'onlineSavingsMsg'
Learn More

3-Allyloxy-1,2-propanediol 99.0+%, TCI America™

Catalog No. G018925G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G018925G 25 g
G0189500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. G018925G Supplier TCI America Supplier No. G018925G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 123-34-2
Molecular Formula C6H12O3
Molecular Weight (g/mol) 132.159
MDL Number MFCD00004721
InChI Key PAKCOSURAUIXFG-UHFFFAOYSA-N
Synonym 3-allyloxy-1,2-propanediol, 3-allyloxy propane-1,2-diol, glycerol 1-allylether, ether, allyl glyceryl, 3-allyloxy propanediol, 1-allyloxy-2,3-propanediol, glycerin 1-allyl ether, glycerol alpha-allyl ether, glycerol alpha-monoallyl ether, 3-2-propenyloxy-1,2-propanediol
PubChem CID 78950
IUPAC Name 3-prop-2-enoxypropane-1,2-diol
SMILES C=CCOCC(CO)O

Specifications

Color Yellow
pH 6.5 tp 7.5 (50% H2O 20°C)
Boiling Point 142°C
Quantity 25 g
Formula Weight 132.16
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Allyloxy-1,2-propanediol
EINECSNumber (2)-0416
RTECSNumber TY2700000
TSCA Yes