missing translation for 'onlineSavingsMsg'
Learn More

3-(Acryloyloxy)propyltrimethoxysilane, 94%, stab. with 100ppm BHT

Catalog No. p-7046808
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1640006 5 g
AAL1640014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1640006 Supplier Thermo Scientific Chemicals Supplier No. L1640006
Only null left

CAS: 4369-14-6 | C9H18O5Si | 234.32 g/mol

3-(Acryloyloxy)propyltrimethoxysilane is used to enhance the thermal conductivity of silicone rubber filled with hybrid fillers. It is involved in the synthesis of vinyl and acryalte modified silica nanoparticles by using vinyltrimethoxysilane. Further, it is used as a silane coupling agent involved in the modification of fluorescein isothiocyanate-mesoporous silica nanoparticles with carbon-carbon double bonds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-(Acryloyloxy)propyltrimethoxysilane is used to enhance the thermal conductivity of silicone rubber filled with hybrid fillers. It is involved in the synthesis of vinyl and acryalte modified silica nanoparticles by using vinyltrimethoxysilane. Further, it is used as a silane coupling agent involved in the modification of fluorescein isothiocyanate-mesoporous silica nanoparticles with carbon-carbon double bonds.

Solubility
Miscible with organic solvents.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents, acids and bases. Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place.

Chemical Identifiers

CAS 4369-14-6
Molecular Formula C9H18O5Si
Molecular Weight (g/mol) 234.32
MDL Number MFCD00054803
InChI Key KBQVDAIIQCXKPI-UHFFFAOYSA-N
Synonym 3-trimethoxysilyl propyl acrylate, 3-acryloyloxy propyltrimethoxysilane, 3-acryloxypropyltrimethoxysilane, 2-propenoic acid, 3-trimethoxysilyl propyl ester, 3-acryloxypropyl trimethoxysilane, 3-trimethoxysilyl propyl 2-propenoate, 3-acryloyloxypropyltrimethoxysilane, 3-acryloxypropyltrimethoxysilane, stab. with 100ppm bht, acmc-1aoz6, acrylic acid, 3-trimethoxysilyl propyl ester
PubChem CID 151204
IUPAC Name 3-trimethoxysilylpropyl prop-2-enoate
SMILES CO[Si](CCCOC(=O)C=C)(OC)OC

Specifications

Boiling Point 95°C to 100°C (3mmHg)
Quantity 5 g
Linear Formula CH2=CHCO2(CH2)3Si(OCH3)3
UN Number UN1760
Beilstein 2046320
Flash Point 123°C (253°F)
Solubility Information Miscible with organic solvents.
Formula Weight 234.33
Percent Purity 94%
Refractive Index 1.429
Sensitivity Moisture sensitive
Density 1.059
Chemical Name or Material 3-(Acryloyloxy)propyltrimethoxysilane, stabilized with 100ppm BHT
Show More Show Less
Hazard Category H314-H317-H332-H335
Hazard Statement GHS H Statement
H314-H332-H317
Causes severe skin burns and eye damage.
Harmful if inhaled.
May cause an allergic skin reaction.
Precautionary Statement P260-P264b-P271-P272-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P333+P313-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, N.O.S.
EINECSNumber 419-560-6
RTECSNumber UD3810000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only