missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Acetylthiophene 95.0+%, TCI America™
$214.37 - $214.37
Chemical Identifiers
| CAS | 1468-83-3 |
|---|---|
| Molecular Formula | C6H6OS |
| Molecular Weight (g/mol) | 126.17 |
| MDL Number | MFCD00005468 |
| InChI Key | RNIDWJDZNNVFDY-UHFFFAOYSA-N |
| Synonym | 3-acetylthiophene, methyl 3-thienyl ketone, 1-thiophen-3-yl ethan-1-one, 1-thiophen-3-yl ethanone, 1-3-thienyl ethanone, ethanone, 1-3-thienyl, ketone, methyl 3-thienyl, 3-acetyl thiophene, 1-thien-3-ylethanone, methyl-3-thienyl ketone |
| PubChem CID | 15116 |
| IUPAC Name | 1-(thiophen-3-yl)ethan-1-one |
| SMILES | CC(=O)C1=CSC=C1 |
Chemical Identifiers
| 1468-83-3 | |
| 126.17 | |
| RNIDWJDZNNVFDY-UHFFFAOYSA-N | |
| 15116 | |
| CC(=O)C1=CSC=C1 |
| C6H6OS | |
| MFCD00005468 | |
| 3-acetylthiophene, methyl 3-thienyl ketone, 1-thiophen-3-yl ethan-1-one, 1-thiophen-3-yl ethanone, 1-3-thienyl ethanone, ethanone, 1-3-thienyl, ketone, methyl 3-thienyl, 3-acetyl thiophene, 1-thien-3-ylethanone, methyl-3-thienyl ketone | |
| 1-(thiophen-3-yl)ethan-1-one |
Specifications
| 1468-83-3 | |
| 95°C | |
| MFCD00005468 | |
| 3-acetylthiophene, methyl 3-thienyl ketone, 1-thiophen-3-yl ethan-1-one, 1-thiophen-3-yl ethanone, 1-3-thienyl ethanone, ethanone, 1-3-thienyl, ketone, methyl 3-thienyl, 3-acetyl thiophene, 1-thien-3-ylethanone, methyl-3-thienyl ketone | |
| CC(=O)C1=CSC=C1 | |
| 126.17 | |
| 126.17 | |
| Crystalline Powder |
| 60°C | |
| C6H6OS | |
| 10 g | |
| RNIDWJDZNNVFDY-UHFFFAOYSA-N | |
| 1-(thiophen-3-yl)ethan-1-one | |
| 15116 | |
| ≥95.0% (GC) | |
| 3-Acetylthiophene |
Safety and Handling
TSCA : No