missing translation for 'onlineSavingsMsg'
Learn More

3-Acetylthiophene 95.0+%, TCI America™

Catalog No. A100610G
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A100610G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A100610G Supplier TCI America Supplier No. A100610G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1468-83-3
Molecular Formula C6H6OS
Molecular Weight (g/mol) 126.17
MDL Number MFCD00005468
InChI Key RNIDWJDZNNVFDY-UHFFFAOYSA-N
Synonym 3-acetylthiophene, methyl 3-thienyl ketone, 1-thiophen-3-yl ethan-1-one, 1-thiophen-3-yl ethanone, 1-3-thienyl ethanone, ethanone, 1-3-thienyl, ketone, methyl 3-thienyl, 3-acetyl thiophene, 1-thien-3-ylethanone, methyl-3-thienyl ketone
PubChem CID 15116
IUPAC Name 1-(thiophen-3-yl)ethan-1-one
SMILES CC(=O)C1=CSC=C1

Specifications

Melting Point 60°C
Boiling Point 95°C
Quantity 10 g
Formula Weight 126.17
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Acetylthiophene