missing translation for 'onlineSavingsMsg'
Learn More

3-Acetylthiophene, 98%

Catalog No. AAA1107830
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1107830 250 g
AAA1107809 10 g
AAA1107818 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1107830 Supplier Thermo Scientific Chemicals Supplier No. A1107830

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1468-83-3 | C6H6OS | 126.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1468-83-3
Molecular Formula C6H6OS
Molecular Weight (g/mol) 126.17
MDL Number MFCD00005468
InChI Key RNIDWJDZNNVFDY-UHFFFAOYSA-N
Synonym 3-acetylthiophene, methyl 3-thienyl ketone, 1-thiophen-3-yl ethan-1-one, 1-thiophen-3-yl ethanone, 1-3-thienyl ethanone, ethanone, 1-3-thienyl, ketone, methyl 3-thienyl, 3-acetyl thiophene, 1-thien-3-ylethanone, methyl-3-thienyl ketone
PubChem CID 15116
IUPAC Name 1-thiophen-3-ylethanone
SMILES CC(=O)C1=CSC=C1

Specifications

Melting Point 58°C to 61°C
Boiling Point 208°C to 210°C
Quantity 250 g
UN Number UN2811
Beilstein 107241
Sensitivity Light sensitive
Formula Weight 126.18
Percent Purity 98%
Chemical Name or Material 3-Acetylthiophene
Hazard Category H302+H312+H332
Hazard Statement GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 215-996-4
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only