missing translation for 'onlineSavingsMsg'
Learn More

3-Acetylpyrrole, 97%

Catalog No. AAA1207303
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1207303 1 g
AAA1207306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1207303 Supplier Thermo Scientific Chemicals Supplier No. A1207303
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1072-82-8 | C6H7NO | 109.128 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1072-82-8
Molecular Formula C6H7NO
Molecular Weight (g/mol) 109.128
MDL Number MFCD00067759
InChI Key KHHSXHXUQVNBGA-UHFFFAOYSA-N
Synonym 3-acetylpyrrole, 1-1h-pyrrol-3-yl ethanone, 1-1h-pyrrol-3-yl ethan-1-one, ethanone, 1-1h-pyrrol-3-yl, 1-1h-pyrrol-3-yl-ethanone, 3-acetyl-1h-pyrrole, 3-acetyl pyrrole, pubchem22508, 3-acetyl-1h-pyrroline
PubChem CID 2737793
IUPAC Name 1-(1H-pyrrol-3-yl)ethanone
SMILES CC(=O)C1=CNC=C1

Specifications

Melting Point 110°C to 114°C
Odor Odorless
Quantity 1 g
Beilstein 107151
Formula Weight 109.13
Percent Purity 97%
Chemical Name or Material 3-Acetylpyrrole
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only