missing translation for 'onlineSavingsMsg'
Learn More

3-Acetylpyridine N-Oxide 98.0+%, TCI America™

Catalog No. A29875G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A29875G 5 g
A29871G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A29875G Supplier TCI America Supplier No. A29875G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 14188-94-4
Molecular Formula C7H7NO2
Molecular Weight (g/mol) 137.138
MDL Number MFCD00955798
InChI Key AZFMFGIMDGBJRR-UHFFFAOYSA-N
PubChem CID 123338
IUPAC Name 1-(1-oxidopyridin-1-ium-3-yl)ethanone
SMILES CC(=O)C1=C[N+](=CC=C1)[O-]

Specifications

Melting Point 146°C
Quantity 5 g
Formula Weight 137.14
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Acetylpyridine N-Oxide