missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Acetylpyridine adenine dinucleotide, 90%, Thermo Scientific™
Chemical Identifiers
| CAS | 86-08-8 |
|---|---|
| Molecular Formula | C22H28N6O14P2 |
| Molecular Weight (g/mol) | 662.45 |
| InChI Key | KPVQNXLUPNWQHM-RBEMOOQDSA-N |
| Synonym | 3-acetylpyridine adenine dinucleotide, acpyad, 3-acetyl nad, 3-acetylpyridine-dpn, 3-acetylpyridine ad, dpn 3-acetylpyridine analog, 3-acetylpyridine analog of dpn, acetylpyridine adenine dinucleotide, acetylpyridine-adenine dinucleotide, 3-acetylpyridineadenine dinucleotide |
| PubChem CID | 123926 |
| IUPAC Name | [(2R,3S,4R,5R)-5-(3-acetylpyridin-1-ium-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl [[(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy-hydroxyphosphoryl] phosphate |
| SMILES | CC(=O)C1=C[N+](=CC=C1)C2C(C(C(O2)COP(=O)([O-])OP(=O)(O)OCC3C(C(C(O3)N4C=NC5=C4N=CN=C5N)O)O)O)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AC457682500
|
Thermo Scientific Chemicals
457682500 |
250mg |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC457680010
|
Thermo Scientific Chemicals
457680010 |
1g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 86-08-8 | |
| 662.45 | |
| 3-acetylpyridine adenine dinucleotide, acpyad, 3-acetyl nad, 3-acetylpyridine-dpn, 3-acetylpyridine ad, dpn 3-acetylpyridine analog, 3-acetylpyridine analog of dpn, acetylpyridine adenine dinucleotide, acetylpyridine-adenine dinucleotide, 3-acetylpyridineadenine dinucleotide | |
| [(2R,3S,4R,5R)-5-(3-acetylpyridin-1-ium-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl [[(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy-hydroxyphosphoryl] phosphate |
| C22H28N6O14P2 | |
| KPVQNXLUPNWQHM-RBEMOOQDSA-N | |
| 123926 | |
| CC(=O)C1=C[N+](=CC=C1)C2C(C(C(O2)COP(=O)([O-])OP(=O)(O)OCC3C(C(C(O3)N4C=NC5=C4N=CN=C5N)O)O)O)O |
Specifications
| Authentic | |
| C22H28N6O14P2 | |
| 3-acetylpyridine adenine dinucleotide, acpyad, 3-acetyl nad, 3-acetylpyridine-dpn, 3-acetylpyridine ad, dpn 3-acetylpyridine analog, 3-acetylpyridine analog of dpn, acetylpyridine adenine dinucleotide, acetylpyridine-adenine dinucleotide, 3-acetylpyridineadenine dinucleotide | |
| CC(=O)C1=C[N+](=CC=C1)C2C(C(C(O2)COP(=O)([O-])OP(=O)(O)OCC3C(C(C(O3)N4C=NC5=C4N=CN=C5N)O)O)O)O | |
| 662.45 | |
| 662.45 | |
| 3-Acetylpyridine adenine dinucleotide |
| 89% min. (HPLC) | |
| 250mg | |
| KPVQNXLUPNWQHM-RBEMOOQDSA-N | |
| [(2R,3S,4R,5R)-5-(3-acetylpyridin-1-ium-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl [[(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy-hydroxyphosphoryl] phosphate | |
| 123926 | |
| 90% |
Safety and Handling
EINECSNumber : 201-649-4
RUO â Research Use Only