missing translation for 'onlineSavingsMsg'
Learn More

3-Acetylpyridine adenine dinucleotide, 90%

Catalog No. AC457682500
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC457682500 250 mg
AC457680010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC457682500 Supplier Thermo Scientific Chemicals Supplier No. 457682500
Only null left

CAS: 86-08-8 | C22H28N6O14P2 | 662.44 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 86-08-8
Molecular Formula C22H28N6O14P2
Molecular Weight (g/mol) 662.44
MDL Number MFCD00078882
InChI Key KPVQNXLUPNWQHM-RBEMOOQDSA-N
Synonym 3-acetylpyridine adenine dinucleotide, acpyad, 3-acetyl nad, 3-acetylpyridine-dpn, 3-acetylpyridine ad, dpn 3-acetylpyridine analog, 3-acetylpyridine analog of dpn, acetylpyridine adenine dinucleotide, acetylpyridine-adenine dinucleotide, 3-acetylpyridineadenine dinucleotide
PubChem CID 123926
SMILES CC(=O)C1=CC=C[N+](=C1)[C@@H]1O[C@H](COP([O-])(=O)OP(O)(=O)OC[C@H]2O[C@H]([C@H](O)[C@@H]2O)N2C=NC3=C(N)N=CN=C23)[C@@H](O)[C@H]1O

Specifications

Infrared Spectrum Authentic
Assay Percent Range 89% min. (HPLC)
Quantity 250 mg
IUPAC Name 3-acetyl-1-[(2R,3R,4S,5R)-5-{[({[(2R,3S,4R,5R)-5-(6-amino-9H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy}(hydroxy)phosphoryl phosphonato)oxy]methyl}-3,4-dihydroxyoxolan-2-yl]-1λ⁵-pyridin-1-ylium
Formula Weight 662.45
Percent Purity 90%
Chemical Name or Material 3-Acetylpyridine adenine dinucleotide
EINECSNumber 201-649-4

RUO – Research Use Only