missing translation for 'onlineSavingsMsg'
Learn More

3-Acetylphenyl isothiocyanate, 97%

Catalog No. AAL1013403
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1013403 1 g
AAL1013406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1013403 Supplier Thermo Scientific Chemicals Supplier No. L1013403

CAS: 3125-71-1 | C9H7NOS | 177.221 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3125-71-1
Molecular Formula C9H7NOS
Molecular Weight (g/mol) 177.221
MDL Number MFCD00041081
InChI Key RGANVWCPAAIVNN-UHFFFAOYSA-N
Synonym 3-acetylphenyl isothiocyanate, 1-3-isothiocyanatophenyl ethanone, 3-acetylphenylisothiocyanate, 1-3-isothiocyanato-phenyl-ethanone, 3-acetylbenzenisothiocyanate, acmc-20anak, 3'-isothiocyanatoacetophenone, 3-methylcarbonylphenyl isothiocyanate, 1-3-isothiocyanatophenyl ethanone #, ethanone,1-3-isothiocyanatophenyl
PubChem CID 520515
IUPAC Name 1-(3-isothiocyanatophenyl)ethanone
SMILES CC(=O)C1=CC(=CC=C1)N=C=S

Specifications

Melting Point ∼24°C
Boiling Point 135°C to 136°C (2 mmHg)
Flash Point >110°C (230°F)
Odor Irritating
Refractive Index 1.648
Quantity 1 g
UN Number UN2923
Beilstein 2803021
Sensitivity Moisture sensitive
Formula Weight 177.23
Percent Purity 97%
Chemical Name or Material 3-Acetylphenyl isothiocyanate
Show More Show Less
Hazard Category H302+H312-H314-H331-H335
Hazard Statement GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only