missing translation for 'onlineSavingsMsg'
Learn More

3-Acetylindole, 98%

CAS: 703-80-0 | C10H9NO | 159.188 g/mol

Supplier:  Thermo Scientific Chemicals A1622114

 View more versions of this product

Catalog No. AAA1622114


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

3-Acetylindole
186°C to 192°C
MFCD00005626
122579
VUIMBZIZZFSQEE-UHFFFAOYSA-N
1-(1H-indol-3-yl)ethanone
12802
98%
703-80-0
C10H9NO
25 g
3-acetylindole, 1-1h-indol-3-yl ethanone, 3-acetyl-1h-indole, ethanone, 1-1h-indol-3-yl, acetyl-3-indole, 1-1h-indol-3-yl-ethanone, ketone, indol-3-yl methyl, 3-acetyl indole, indol-3-yl methyl ketone, 1-1h-indol-3-yl ethan-1-one
CC(=O)C1=CNC2=CC=CC=C21
159.188
159.19

Chemical Identifiers

703-80-0
159.188
VUIMBZIZZFSQEE-UHFFFAOYSA-N
12802
CC(=O)C1=CNC2=CC=CC=C21
C10H9NO
MFCD00005626
3-acetylindole, 1-1h-indol-3-yl ethanone, 3-acetyl-1h-indole, ethanone, 1-1h-indol-3-yl, acetyl-3-indole, 1-1h-indol-3-yl-ethanone, ketone, indol-3-yl methyl, 3-acetyl indole, indol-3-yl methyl ketone, 1-1h-indol-3-yl ethan-1-one
1-(1H-indol-3-yl)ethanone
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 211-875-5

RTECSNumber : OB4520000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only