missing translation for 'onlineSavingsMsg'
Learn More

3-Acetylindole, 98%

Catalog No. AAA1622122
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1622122 100 g
AAA1622106 5 g
AAA1622114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1622122 Supplier Thermo Scientific Chemicals Supplier No. A1622122

CAS: 703-80-0 | C10H9NO | 159.188 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 703-80-0
Molecular Formula C10H9NO
Molecular Weight (g/mol) 159.188
MDL Number MFCD00005626
InChI Key VUIMBZIZZFSQEE-UHFFFAOYSA-N
Synonym 3-acetylindole, 1-1h-indol-3-yl ethanone, 3-acetyl-1h-indole, ethanone, 1-1h-indol-3-yl, acetyl-3-indole, 1-1h-indol-3-yl-ethanone, ketone, indol-3-yl methyl, 3-acetyl indole, indol-3-yl methyl ketone, 1-1h-indol-3-yl ethan-1-one
PubChem CID 12802
IUPAC Name 1-(1H-indol-3-yl)ethanone
SMILES CC(=O)C1=CNC2=CC=CC=C21

Specifications

Melting Point 186°C to 192°C
Quantity 100 g
Beilstein 122579
Formula Weight 159.19
Percent Purity 98%
Chemical Name or Material 3-Acetylindole
EINECSNumber 211-875-5
RTECSNumber OB4520000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only