missing translation for 'onlineSavingsMsg'
Learn More

3-Acetylindole, 97%

CAS: 703-80-0 | C10H9NO | 159.19 g/mol

$423.34 - $423.34

Chemical Identifiers

CAS 703-80-0
Molecular Formula C10H9NO
Molecular Weight (g/mol) 159.19
MDL Number MFCD00005626
InChI Key VUIMBZIZZFSQEE-UHFFFAOYSA-N
Synonym 3-acetylindole, 1-1h-indol-3-yl ethanone, 3-acetyl-1h-indole, ethanone, 1-1h-indol-3-yl, acetyl-3-indole, 1-1h-indol-3-yl-ethanone, ketone, indol-3-yl methyl, 3-acetyl indole, indol-3-yl methyl ketone, 1-1h-indol-3-yl ethan-1-one
PubChem CID 12802
IUPAC Name 1-(1H-indol-3-yl)ethanone
SMILES CC(=O)C1=CNC2=CC=CC=C21
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC102590250
SDS View Documents
Thermo Scientific Chemicals
102590250
25 g Glass bottle
Each for $423.34
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

703-80-0
159.19
VUIMBZIZZFSQEE-UHFFFAOYSA-N
12802
CC(=O)C1=CNC2=CC=CC=C21
C10H9NO
MFCD00005626
3-acetylindole, 1-1h-indol-3-yl ethanone, 3-acetyl-1h-indole, ethanone, 1-1h-indol-3-yl, acetyl-3-indole, 1-1h-indol-3-yl-ethanone, ketone, indol-3-yl methyl, 3-acetyl indole, indol-3-yl methyl ketone, 1-1h-indol-3-yl ethan-1-one
1-(1H-indol-3-yl)ethanone

Specifications

703-80-0
100.0
Yellow to Brown
97%
C10H9NO
25 g
3-acetylindole, 1-1h-indol-3-yl ethanone, 3-acetyl-1h-indole, ethanone, 1-1h-indol-3-yl, acetyl-3-indole, 1-1h-indol-3-yl-ethanone, ketone, indol-3-yl methyl, 3-acetyl indole, indol-3-yl methyl ketone, 1-1h-indol-3-yl ethan-1-one
CC(=O)C1=CNC2=CC=CC=C21
159.19
159.19
Powder
96.0
185°C to 191°C
Authentic
Glass bottle
MFCD00005626
21, 316
VUIMBZIZZFSQEE-UHFFFAOYSA-N
1-(1H-indol-3-yl)ethanone
12802
97%
3-Acetylindole, 97%
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 211-875-5

RTECSNumber : OB4520000

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only