missing translation for 'onlineSavingsMsg'
Learn More

3-Acetylindole, 97%

Catalog No. AC102590050
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC102590050 5 g Glass bottle
AC102590250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC102590050 Supplier Thermo Scientific Chemicals Supplier No. 102590050

CAS: 703-80-0 | C10H9NO | 159.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 703-80-0
Molecular Formula C10H9NO
Molecular Weight (g/mol) 159.19
MDL Number MFCD00005626
InChI Key VUIMBZIZZFSQEE-UHFFFAOYSA-N
Synonym 3-acetylindole, 1-1h-indol-3-yl ethanone, 3-acetyl-1h-indole, ethanone, 1-1h-indol-3-yl, acetyl-3-indole, 1-1h-indol-3-yl-ethanone, ketone, indol-3-yl methyl, 3-acetyl indole, indol-3-yl methyl ketone, 1-1h-indol-3-yl ethan-1-one
PubChem CID 12802
IUPAC Name 1-(1H-indol-3-yl)ethanone
SMILES CC(=O)C1=CNC2=CC=CC=C21

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 185°C to 191°C
Color Yellow to Brown
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 5 g
Beilstein 21, 316
Formula Weight 159.19
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 3-Acetylindole, 97%
Show More Show Less
EINECSNumber 211-875-5
RTECSNumber OB4520000

RUO – Research Use Only