missing translation for 'onlineSavingsMsg'
Learn More

3-Acetylbenzeneboronic acid, 97%

Catalog No. AAB2347803
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2347803 1 g
AAB2347806 5 g
AAB2347814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2347803 Supplier Thermo Scientific Chemicals Supplier No. B2347803
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 204841-19-0 | C8H9BO3 | 163.967 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 204841-19-0
Molecular Formula C8H9BO3
Molecular Weight (g/mol) 163.967
MDL Number MFCD01074678
InChI Key SJGGDZCTGBKBCK-UHFFFAOYSA-N
Synonym 3-acetylphenyl boronic acid, 3-acetylbenzeneboronic acid, 3-acetylphenylboronicacid, m-acetylphenylboronic acid, 3'-boronoacetophenone, boronic acid, 3-acetylphenyl, 1-3-dihydroxyboranyl phenyl ethan-1-one, pubchem2751, acmc-209faq, 3-acetylphenyboronic acid
PubChem CID 2734310
ChEBI CHEBI:86552
IUPAC Name (3-acetylphenyl)boronic acid
SMILES B(C1=CC(=CC=C1)C(=O)C)(O)O

Specifications

Melting Point ∼200°C to 205°C
Quantity 1 g
Beilstein 8255673
Formula Weight 163.97
Percent Purity 97%
Chemical Name or Material 3-Acetylbenzeneboronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only