missing translation for 'onlineSavingsMsg'
Learn More

3-Acetyl-6-bromopyridine, 97%

Catalog No. AAH3320406
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3320406 5 g
AAH3320403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3320406 Supplier Thermo Scientific Chemicals Supplier No. H3320406

CAS: 139042-59-4 | C7H6BrNO | 200.035 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 139042-59-4
Molecular Formula C7H6BrNO
Molecular Weight (g/mol) 200.035
MDL Number MFCD04974527
InChI Key MUKKGHQBUKOMTD-UHFFFAOYSA-N
Synonym 5-acetyl-2-bromopyridine, 1-6-bromopyridin-3-yl ethanone, 2-bromo-5-acetylpyridine, 1-6-bromo-pyridin-3-yl-ethanone, 1-6-bromopyridin-3-yl ethan-1-one, 3-acetyl-6-bromopyridine, ethanone, 1-6-bromo-3-pyridinyl, 1-6-bromopyrid-3-yl ethanone, 1-6-bromo-3-pyridyl ethanone, acmc-209cij
PubChem CID 15668195
IUPAC Name 1-(6-bromopyridin-3-yl)ethanone
SMILES CC(=O)C1=CN=C(C=C1)Br

Specifications

Melting Point 124°C to 128°C
Quantity 5 g
Formula Weight 200.03
Percent Purity 97%
Chemical Name or Material 3-Acetyl-6-bromopyridine
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only