missing translation for 'onlineSavingsMsg'
Learn More

3-Acetyl-6-bromocoumarin 96.0+%, TCI America™

Catalog No. A27231G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A27231G 1 g
A27235G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A27231G Supplier TCI America Supplier No. A27231G
Only null left

Chemical Identifiers

CAS 2199-93-1
Molecular Formula C11H7BrO3
Molecular Weight (g/mol) 267.08
MDL Number MFCD00024075
InChI Key XFQYOFLFNKCHLG-UHFFFAOYSA-N
Synonym 3-acetyl-6-bromocoumarin, 3-acetyl-6-bromo-2h-chromen-2-one, 3-acetyl-6-bromo-chromen-2-one, 2h-1-benzopyran-2-one,3-acetyl-6-bromo, 3-acetyl-6-bromo-2-oxochromene, pubchem13336, maybridge1_000212, zerenex e/1034014, 6-bromanyl-3-ethanoyl-chromen-2-one
PubChem CID 304813
IUPAC Name 3-acetyl-6-bromo-2H-chromen-2-one
SMILES CC(=O)C1=CC2=CC(Br)=CC=C2OC1=O

Specifications

Melting Point 225°C
Quantity 1 g
Formula Weight 267.08
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Acetyl-6-bromocoumarin