missing translation for 'onlineSavingsMsg'
Learn More

3-Acetyl-4-methylpyridine, 96%

CAS: 51227-30-6 | C8H9NO | 135.166 g/mol

$128.34 - $414.85

Chemical Identifiers

CAS 51227-30-6
Molecular Formula C8H9NO
Molecular Weight (g/mol) 135.166
MDL Number MFCD08447082
InChI Key LCEINQHGZAFADC-UHFFFAOYSA-N
Synonym 1-4-methylpyridin-3-yl ethanone, 3-acetyl-4-methylpyridine, 1-4-methyl-3-pyridinyl ethanone, 1-4-methyl-3-pyridinyl-ethanone, 1-4-methylpyridin-3-yl ethan-1-one, 1-4-methyl-pyridin-3-yl-ethanone, ethanone, 1-4-methyl-3-pyridinyl, 4-methyl-3-acetylpyridine, 3-acetyl-4-picoline, methyl 4-picolyl ketone
PubChem CID 10942481
IUPAC Name 1-(4-methylpyridin-3-yl)ethanone
SMILES CC1=C(C=NC=C1)C(=O)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH3338003
View Documents
Thermo Scientific Chemicals
H3338003
1 g
Each for $128.34
Only null left
 
AAH3338006
View Documents
Thermo Scientific Chemicals
H3338006
5 g
Each for $414.85
Only null left
 
Description

Description

It is used as pharmaceutical intermediate. Benzoylthiophenes are allosteric enhancers (AE) of agonist activity at the A1 adenosine receptor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as pharmaceutical intermediate. Benzoylthiophenes are allosteric enhancers (AE) of agonist activity at the A1 adenosine receptor.

Solubility
Sparingly soluble in water.(0.26 g/L) (25°C),

Notes
Store in cool dry place. Ensure proper ventilation. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

51227-30-6
135.166
LCEINQHGZAFADC-UHFFFAOYSA-N
10942481
CC1=C(C=NC=C1)C(=O)C
C8H9NO
MFCD08447082
1-4-methylpyridin-3-yl ethanone, 3-acetyl-4-methylpyridine, 1-4-methyl-3-pyridinyl ethanone, 1-4-methyl-3-pyridinyl-ethanone, 1-4-methylpyridin-3-yl ethan-1-one, 1-4-methyl-pyridin-3-yl-ethanone, ethanone, 1-4-methyl-3-pyridinyl, 4-methyl-3-acetylpyridine, 3-acetyl-4-picoline, methyl 4-picolyl ketone
1-(4-methylpyridin-3-yl)ethanone

Specifications

51227-30-6
C8H9NO
MFCD08447082
1-4-methylpyridin-3-yl ethanone, 3-acetyl-4-methylpyridine, 1-4-methyl-3-pyridinyl ethanone, 1-4-methyl-3-pyridinyl-ethanone, 1-4-methylpyridin-3-yl ethan-1-one, 1-4-methyl-pyridin-3-yl-ethanone, ethanone, 1-4-methyl-3-pyridinyl, 4-methyl-3-acetylpyridine, 3-acetyl-4-picoline, methyl 4-picolyl ketone
LCEINQHGZAFADC-UHFFFAOYSA-N
1-(4-methylpyridin-3-yl)ethanone
10942481
96%
57°C to 58°C (1 mmHg)
1.5295
1 g
Sparingly soluble in water.(0.26g/L) (25°C),
CC1=C(C=NC=C1)C(=O)C
135.166
135.17
3-Acetyl-4-methylpyridine
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only