missing translation for 'onlineSavingsMsg'
Learn More

3-Acetyl-4-methylpyridine, 96%

Catalog No. AAH3338006
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3338006 5 g
AAH3338003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3338006 Supplier Thermo Scientific Chemicals Supplier No. H3338006
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 51227-30-6 | C8H9NO | 135.166 g/mol

It is used as pharmaceutical intermediate. Benzoylthiophenes are allosteric enhancers (AE) of agonist activity at the A1 adenosine receptor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as pharmaceutical intermediate. Benzoylthiophenes are allosteric enhancers (AE) of agonist activity at the A1 adenosine receptor.

Solubility
Sparingly soluble in water.(0.26 g/L) (25°C),

Notes
Store in cool dry place. Ensure proper ventilation. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 51227-30-6
Molecular Formula C8H9NO
Molecular Weight (g/mol) 135.166
MDL Number MFCD08447082
InChI Key LCEINQHGZAFADC-UHFFFAOYSA-N
Synonym 1-4-methylpyridin-3-yl ethanone, 3-acetyl-4-methylpyridine, 1-4-methyl-3-pyridinyl ethanone, 1-4-methyl-3-pyridinyl-ethanone, 1-4-methylpyridin-3-yl ethan-1-one, 1-4-methyl-pyridin-3-yl-ethanone, ethanone, 1-4-methyl-3-pyridinyl, 4-methyl-3-acetylpyridine, 3-acetyl-4-picoline, methyl 4-picolyl ketone
PubChem CID 10942481
IUPAC Name 1-(4-methylpyridin-3-yl)ethanone
SMILES CC1=C(C=NC=C1)C(=O)C

Specifications

Boiling Point 57°C to 58°C (1 mmHg)
Refractive Index 1.5295
Quantity 5 g
Solubility Information Sparingly soluble in water.(0.26g/L) (25°C),
Formula Weight 135.17
Percent Purity 96%
Chemical Name or Material 3-Acetyl-4-methylpyridine
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only