missing translation for 'onlineSavingsMsg'
Learn More

3-Acetyl-2-methyl-5-phenylthiophene, 98%, Thermo Scientific Chemicals

Catalog No. AAL1355406
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1355406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1355406 Supplier Thermo Scientific Chemicals Supplier No. L1355406
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 40932-63-6 | Molecular Formula: C13H12OS | Molecular Weight: 216.30 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 40932-63-6
Molecular Formula C13H12OS
Molecular Weight (g/mol) 216.30
MDL Number MFCD00151790
InChI Key GREAZYFTAJMZFD-UHFFFAOYSA-N
Synonym 3-acetyl-2-methyl-5-phenylthiophene, 1-2-methyl-5-phenylthiophen-3-yl ethanone, 1-2-methyl-5-phenylthien-3-yl ethanone, ethanone, 1-2-methyl-5-phenyl-3-thienyl, acmc-20anao, 3-acetyl-2-methyl-5-phenylthiohene, 1-2-methyl-5-phenyl-3-thienyl ethanone, 1-2-methyl-5-phenyl-3-thiophenyl ethanone, 1-2-methyl-5-phenyl-thiophen-3-yl ethanone, 1-2-methyl-5-phenyl-thiophen-3-yl-ethanone
PubChem CID 2728765
IUPAC Name 1-(2-methyl-5-phenylthiophen-3-yl)ethanone
SMILES CC(=O)C1=C(C)SC(=C1)C1=CC=CC=C1

Specifications

Melting Point 67°C to 69°C
Quantity 5 g
Beilstein 8481276
Formula Weight 216.3
Percent Purity 98%
Chemical Name or Material 3-Acetyl-2-methyl-5-phenylthiophene
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only