missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Acetyl-2-(aminosulfonyl)-5-chlorothiophene 98.0+%, TCI America™
$158.44 - $548.03
Chemical Identifiers
| CAS | 160982-10-5 |
|---|---|
| Molecular Formula | C6H6ClNO3S2 |
| Molecular Weight (g/mol) | 239.69 |
| MDL Number | MFCD09033309 |
| InChI Key | ODLFFSHLXVZFPY-UHFFFAOYSA-N |
| Synonym | 3-Acetyl-5-chloro-2-thiophenesulfonamide |
| PubChem CID | 15294528 |
| IUPAC Name | 3-acetyl-5-chlorothiophene-2-sulfonamide |
| SMILES | CC(=O)C1=C(SC(Cl)=C1)S(N)(=O)=O |
Chemical Identifiers
| 160982-10-5 | |
| 239.69 | |
| ODLFFSHLXVZFPY-UHFFFAOYSA-N | |
| 15294528 | |
| CC(=O)C1=C(SC(Cl)=C1)S(N)(=O)=O |
| C6H6ClNO3S2 | |
| MFCD09033309 | |
| 3-Acetyl-5-chloro-2-thiophenesulfonamide | |
| 3-acetyl-5-chlorothiophene-2-sulfonamide |
Specifications
| 160982-10-5 | |
| White-Yellow | |
| MFCD09033309 | |
| 3-Acetyl-5-chloro-2-thiophenesulfonamide | |
| CC(=O)C1=C(SC(Cl)=C1)S(N)(=O)=O | |
| 239.69 | |
| 239.69 | |
| Crystalline Powder |
| 182°C | |
| C6H6ClNO3S2 | |
| 1 g | |
| ODLFFSHLXVZFPY-UHFFFAOYSA-N | |
| 3-acetyl-5-chlorothiophene-2-sulfonamide | |
| 15294528 | |
| ≥98.0% (HPLC,T) | |
| 3-Acetyl-2-(aminosulfonyl)-5-chlorothiophene |
Safety and Handling
TSCA : No