missing translation for 'onlineSavingsMsg'
Learn More

3-Acetyl-2,5-dimethylfuran 98.0+%, TCI America™

Catalog No. a112125g
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A112125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A112125G Supplier TCI America Supplier No. A112125G

Chemical Identifiers

CAS 10599-70-9
Molecular Formula C8H10O2
Molecular Weight (g/mol) 138.166
MDL Number MFCD00010281
InChI Key KBSVBCHYXYXDAG-UHFFFAOYSA-N
Synonym 3-acetyl-2,5-dimethylfuran, 1-2,5-dimethylfuran-3-yl ethanone, ethanone, 1-2,5-dimethyl-3-furanyl, 3-acetyl-2,5-dimethyl furan, 2,5-dimethyl-3-acetylfuran, 1-2,5-dimethyl-3-furyl ethan-1-one, 2,5-dimethyl-3-furyl methyl ketone, 1-2,5-dimethyl-3-furanyl ethanone, 1-2,5-dimethyl-3-furyl ethanone, unii-798v2t7zbv
PubChem CID 61527
IUPAC Name 1-(2,5-dimethylfuran-3-yl)ethanone
SMILES CC1=CC(=C(O1)C)C(=O)C

Specifications

Color Yellow
Boiling Point 196°C
Quantity 25 g
Formula Weight 138.17
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Acetyl-2,5-dimethylfuran