missing translation for 'onlineSavingsMsg'
Learn More

3-Acetyl-2,4-dimethylpyrrole 98.0+%, TCI America™

Catalog No. p-7118047
Change view
Click to view available options
Quantity:
1 g
5 g
Catalog No. Quantity
A27341G 1 g
A27345G 5 g
2 options

Catalog No. A27341G

Supplier: TCI America A27341G

Only null left

Chemical Identifiers

CAS 2386-25-6
Molecular Formula C8H11NO
Molecular Weight (g/mol) 137.182
MDL Number MFCD00005221
InChI Key VGZCKCJMYREIKA-UHFFFAOYSA-N
Synonym 3-acetyl-2,4-dimethylpyrrole, 2,4-dimethyl-3-acetylpyrrole, 1-2,4-dimethyl-1h-pyrrol-3-yl ethanone, 2,4-dimethylpyrrol-3-yl methyl ketone, ethanone, 1-2,4-dimethyl-1h-pyrrol-3-yl, ketone, 2,4-dimethylpyrrol-3-yl methyl, 1-2,4-dimethyl-1h-pyrrol-3-yl ethan-1-one, 2,4-dimethyl-3-acetyl-pyrrole, 3-acetyl-2,5-dimethyl-pyrrole, 1-2,4-dimethyl-1h-pyrrol-3-yl-ethanone
PubChem CID 15163
IUPAC Name 1-(2,4-dimethyl-1H-pyrrol-3-yl)ethanone
SMILES CC1=CNC(=C1C(=O)C)C

Specifications

Melting Point 138°C
Boiling Point 173°C
Quantity 1 g
Formula Weight 137.18
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Acetyl-2,4-dimethylpyrrole
RTECSNumber OB2885000
TSCA No