missing translation for 'onlineSavingsMsg'
Learn More

3-Acetoxy-1-propenylboronic acid pinacol ester, 97%

Catalog No. p-7047744
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL19700MD 250 mg
AAL1970003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL19700MD Supplier Thermo Scientific Chemicals Supplier No. L19700MD
Only null left

CAS: 161395-97-7 | C11H19BO4 | 226.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 161395-97-7
Molecular Formula C11H19BO4
Molecular Weight (g/mol) 226.08
MDL Number MFCD03788751
InChI Key ZXBRLGZFCXGTBA-UHFFFAOYSA-N
Synonym 3-acetoxy-1-propenylboronic acid pinacol ester, e-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl allyl acetate, 2-3-acetoxy-1-propenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2e-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2-propen-1-yl acetate, 2e-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl prop-2-en-1-yl acetate, e-3-acetoxy-1-propenylboronic acid 2,3-dimethylbutane-2,3-diyl ester
PubChem CID 11264613
IUPAC Name [(E)-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)prop-2-enyl] acetate
SMILES CC(=O)OCC=CB1OC(C)(C)C(C)(C)O1

Specifications

Refractive Index 1.4485
Quantity 250 mg
Beilstein 6923397
Formula Weight 226.08
Percent Purity 97%
Chemical Name or Material 3-Acetoxy-1-propenylboronic acid pinacol ester
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only