missing translation for 'onlineSavingsMsg'
Learn More

3-Acetamidotetrahydro-2-thiophenone 98.0+%, TCI America™

Catalog No. p-7117756
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
5 g
Catalog No. Quantity
A21445G 5 g
A214425G 25 g
2 options

Catalog No. A21445G

Supplier: TCI America A21445G

Only null left

Chemical Identifiers

CAS 1195-16-0
Molecular Formula C6H9NO2S
Molecular Weight (g/mol) 159.20
MDL Number MFCD00005480
InChI Key NRFJZTXWLKPZAV-UHFFFAOYNA-N
Synonym citiolone, citiolase, thioxidrene, ahctl, achtl, ahct, dl-n-acetylhomocysteine thiolactone, n-acetylhomocysteine thiolactone, acetamide, n-tetrahydro-2-oxo-3-thienyl, 3-acetamidotetrahydro-2-thiophenone
PubChem CID 14520
IUPAC Name N-(2-oxothiolan-3-yl)acetamide
SMILES CC(=O)NC1CCSC1=O

Specifications

Melting Point 110°C
Color White-Yellow
Quantity 5 g
Formula Weight 159.20
Percent Purity ≥98.0% (GC,N)
Physical Form Crystalline Powder
Chemical Name or Material 3-Acetamidotetrahydro-2-thiophenone
RTECSNumber AC8680000
TSCA Yes