missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Acetamidotetrahydro-2-thiophenone 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 110°C |
| Color | White-Yellow |
| Quantity | 25 g |
| Formula Weight | 159.20 |
| Percent Purity | ≥98.0% (GC,N) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 3-Acetamidotetrahydro-2-thiophenone |
Safety and Handling
| RTECSNumber | AC8680000 |
| TSCA | Yes |